missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole 98.0+%, TCI America™
Supplier: TCI America D38341G
Specifications
| 1-[2-(Di-tert-butylphosphino)phenyl]-3,5-diphenyl-1H-pyrazole | |
| 142°C | |
| C29H33N2P | |
| 1 g | |
| JAIIBHBCNPAPPF-UHFFFAOYSA-N | |
| ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane | |
| 11430600 | |
| ≥98.0% (GC,T) |
| 628333-86-8 | |
| White-Yellow | |
| MFCD09038823 | |
| TrippyPhos | |
| CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C | |
| 440.571 | |
| 440.57 | |
| Crystalline Powder |
Chemical Identifiers
| 628333-86-8 | |
| 440.571 | |
| JAIIBHBCNPAPPF-UHFFFAOYSA-N | |
| 11430600 | |
| CC(C)(C)P(C1=CC=CC=C1N2C(=CC(=N2)C3=CC=CC=C3)C4=CC=CC=C4)C(C)(C)C |
| C29H33N2P | |
| MFCD09038823 | |
| TrippyPhos | |
| ditert-butyl-[2-(3,5-diphenylpyrazol-1-yl)phenyl]phosphane |
Safety and Handling
TSCA : No