accessibility menu, dialog, popup

UserName

CAS RN 628333-86-8

H3CH3CCH3PNNCH3CH3H3C

CAS RN 628333-86-8

IUPAC Name: 1-[2-(di-tert-butylphosphanyl)phenyl]-3,5-diphenyl-1H-pyrazole
Synonyms: 1-[2-(di-tert-butylphosphanyl)phenyl]-3,5-diphenylpyrazole
Molecular Weight (g/mol): 440.57
Molecular Formula: C29H33N2P
InChi Key: JAIIBHBCNPAPPF-UHFFFAOYSA-N
SMILES: CC(C)(C)P(C1=CC=CC=C1N1N=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results