missing translation for 'onlineSavingsMsg'
Learn More

1,2-Di(1-naphthyl)ethane 94.0+%, TCI America™

Catalog No. D43715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43715G 5 g
D43711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43715G Supplier TCI America Supplier No. D43715G

Chemical Identifiers

CAS 15374-45-5
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD00037736
InChI Key OJGSITVFPMSVGU-UHFFFAOYSA-N
Synonym 1,2-di 1-naphthyl ethane, 1,2-bis 1-naphthyl ethane, 1,2-di alpha-naphthyl ethane, ethane, 1,2-di-1-naphthyl, naphthalene, 1,1'-1,2-ethanediyl bis, 1,2-di-.alpha.-naphthylethane, naphthalene,1,1'-1,2-ethanediyl bis, 1-2-naphthalen-1-yl ethyl naphthalene, acmc-1bvox, 1.2-di-a-naphthylathan
PubChem CID 139929
IUPAC Name 1-(2-naphthalen-1-ylethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCC3=CC=CC4=CC=CC=C43

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.39
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Di(1-naphthyl)ethane