CAS RN 15374-45-5
IUPAC Name:
1-[2-(naphthalen-1-yl)ethyl]naphthalene
Synonyms:
1-[2-(naphthalen-1-yl)ethyl]naphthalene
Molecular Weight (g/mol):
282.39
Molecular Formula:
C22H18
InChi Key:
OJGSITVFPMSVGU-UHFFFAOYSA-N
SMILES:
C(CC1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1
1
–
1
of
1
results
1
1,2-Di(1-naphthyl)ethane 94.0+%, TCI America™
CAS: 15374-45-5 Molecular Formula: C22H18 Molecular Weight (g/mol): 282.386 MDL Number: MFCD00037736 InChI Key: OJGSITVFPMSVGU-UHFFFAOYSA-N Synonym: 1,2-di 1-naphthyl ethane,1,2-bis 1-naphthyl ethane,1,2-di alpha-naphthyl ethane,ethane, 1,2-di-1-naphthyl,naphthalene, 1,1'-1,2-ethanediyl bis,1,2-di-.alpha.-naphthylethane,naphthalene,1,1'-1,2-ethanediyl bis,1-2-naphthalen-1-yl ethyl naphthalene,acmc-1bvox,1.2-di-a-naphthylathan PubChem CID: 139929 IUPAC Name: 1-(2-naphthalen-1-ylethyl)naphthalene SMILES: C1=CC=C2C(=C1)C=CC=C2CCC3=CC=CC4=CC=CC=C43