missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Decanediol 99.0+%, TCI America™
$78.29 - $259.11
Chemical Identifiers
| CAS | 1119-86-4 |
|---|---|
| Molecular Formula | C10H22O2 |
| Molecular Weight (g/mol) | 174.284 |
| MDL Number | MFCD00010739 |
| InChI Key | YSRSBDQINUMTIF-UHFFFAOYSA-N |
| Synonym | 1,2-decanediol, decylene glycol, decane 1,2-diol, 1,2-dihydroxydecane, acmc-20apfz, acmc-20apjb, pubchem12680, acmc-1bny3 |
| PubChem CID | 79141 |
| IUPAC Name | decane-1,2-diol |
| SMILES | CCCCCCCCC(CO)O |
Chemical Identifiers
| 1119-86-4 | |
| 174.284 | |
| YSRSBDQINUMTIF-UHFFFAOYSA-N | |
| 79141 | |
| CCCCCCCCC(CO)O |
| C10H22O2 | |
| MFCD00010739 | |
| 1,2-decanediol, decylene glycol, decane 1,2-diol, 1,2-dihydroxydecane, acmc-20apfz, acmc-20apjb, pubchem12680, acmc-1bny3 | |
| decane-1,2-diol |
Specifications
| 1119-86-4 | |
| White | |
| C10H22O2 | |
| 25 g | |
| YSRSBDQINUMTIF-UHFFFAOYSA-N | |
| decane-1,2-diol | |
| 79141 | |
| ≥99.0% (GC) | |
| 1,2-Decanediol |
| 49°C | |
| 168°C | |
| MFCD00010739 | |
| 1,2-decanediol, decylene glycol, decane 1,2-diol, 1,2-dihydroxydecane, acmc-20apfz, acmc-20apjb, pubchem12680, acmc-1bny3 | |
| CCCCCCCCC(CO)O | |
| 174.284 | |
| 174.28 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : No