missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent, 98%, Spectrum™ Chemical
C1466, 125572-95-4, C14H22N2O8·H2O
Supplier: Spectrum Chemical Mfg Cor C1466100GM
Description
Spectrum™ Chemical (1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent is an alkyl-substituted amino acid that functions as a chelating agent. This ingredient′s source is synthetic. It appears as a solid and is partially soluble in cold water. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
| 125572-95-4 | |
| C14H20N2O8 | |
| 100 g | |
| [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O | |
| 344.32 | |
| Reagent |
| 100% | |
| MFCD00149243,MFCD00066429,MFCD00003845 | |
| FCKYPQBAHLOOJQ-NXEZZACHSA-L | |
| 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate | |
| 98% | |
| Amber Glass Bottle |
Chemical Identifiers
| 125572-95-4 | |
| 344.32 | |
| FCKYPQBAHLOOJQ-NXEZZACHSA-L | |
| [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O |
| C14H20N2O8 | |
| MFCD00149243,MFCD00066429,MFCD00003845 | |
| 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate |