missing translation for 'onlineSavingsMsg'
Learn More

(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent, 98%, Spectrum™ Chemical

Catalog No. 18600000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600000 25 g Amber Glass Bottle
18600001 100 g Amber Glass Bottle
18600002 500 g Amber Glass Bottle
18600003 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600000 Supplier Spectrum Chemical Mfg Cor Supplier No. C146625GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1466, 125572-95-4, C14H22N2O8·H2O

Spectrum™ Chemical (1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent is an alkyl-substituted amino acid that functions as a chelating agent. This ingredient′s source is synthetic. It appears as a solid and is partially soluble in cold water. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 125572-95-4
Molecular Formula C14H20N2O8
Molecular Weight (g/mol) 344.32
MDL Number MFCD00149243,MFCD00066429,MFCD00003845
InChI Key FCKYPQBAHLOOJQ-NXEZZACHSA-L
IUPAC Name 2-{[(1R,2R)-2-[bis(carboxylatomethyl)azaniumyl]cyclohexyl](carboxylatomethyl)azaniumyl}acetate
SMILES [O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Quantity 25 g
Packaging Amber Glass Bottle
Percent Purity 98%
Grade Reagent