missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexanedione 97.0+%, TCI America™
Supplier: TCI America C07285G
Specifications
| 1,2-Cyclohexanedione | |
| Yellow | |
| C6H8O2 | |
| 5 g | |
| OILAIQUEIWYQPH-UHFFFAOYSA-N | |
| cyclohexane-1,2-dione | |
| 13006 | |
| 112.13 | |
| Crystals |
| 765-87-7 | |
| 194°C | |
| MFCD00001648 | |
| 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
| C1CCC(=O)C(=O)C1 | |
| 112.128 | |
| CHEBI:41674 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 765-87-7 | |
| 112.128 | |
| OILAIQUEIWYQPH-UHFFFAOYSA-N | |
| 13006 | |
| cyclohexane-1,2-dione |
| C6H8O2 | |
| MFCD00001648 | |
| 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol | |
| CHEBI:41674 | |
| C1CCC(=O)C(=O)C1 |
Safety and Handling
RTECSNumber : GV0340000
TSCA : Yes