missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione 97.0+%, TCI America™

Catalog No. p-7124579
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07285G 5 g
C072825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07285G Supplier TCI America Supplier No. C07285G
Only null left

Chemical Identifiers

CAS 765-87-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001648
InChI Key OILAIQUEIWYQPH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
PubChem CID 13006
ChEBI CHEBI:41674
IUPAC Name cyclohexane-1,2-dione
SMILES C1CCC(=O)C(=O)C1

Specifications

Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 112.13
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 1,2-Cyclohexanedione
RTECSNumber GV0340000
TSCA Yes