missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chlorophenyl)ethanol, 96%

Catalog No. AAB2480014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2480014 25 g
AAB2480006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2480014 Supplier Thermo Scientific Chemicals Supplier No. B2480014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13524-04-4 | C8H9ClO | 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13524-04-4
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00041037
InChI Key DDUBOVLGCYUYFX-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl ethanol, 1-2-chlorophenyl ethan-1-ol, 2-chloro-alpha-methylbenzyl alcohol, 1-2-chlorophenyl-1-ethanol, benzenemethanol, 2-chloro-.alpha.-methyl, benzenemethanol, 2-chloro-alpha-methyl, benzyl alcohol, o-chloro-.alpha.-methyl, 1-2-chloro-phenyl-ethanol, 1-2-chlorophenyl ethyl alcohol, 1-2'-chlorophenyl-1-hydroxyethane
PubChem CID 26082
IUPAC Name 1-(2-chlorophenyl)ethanol
SMILES CC(C1=CC=CC=C1Cl)O

Specifications

Quantity 25 g
Refractive Index 1.545
Boiling Point 121°C to 123°C (14 mmHg)
Formula Weight 156.61
Percent Purity 96%
Chemical Name or Material 1-(2-Chlorophenyl)ethanol
EINECSNumber 236-868-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only