accessibility menu, dialog, popup

UserName

H3COHCl

CAS RN 13524-04-4

IUPAC Name: 1-(2-chlorophenyl)ethan-1-ol
Synonyms: 1-(2-chlorophenyl)ethanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: DDUBOVLGCYUYFX-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)