missing translation for 'onlineSavingsMsg'
Learn More

1,2-Butanediol 98.0+%, TCI America™

Catalog No. B080425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B080425G 25 g
B0804500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B080425G Supplier TCI America Supplier No. B080425G
Only null left

Chemical Identifiers

CAS 584-03-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004570
InChI Key BMRWNKZVCUKKSR-UHFFFAOYNA-N
Synonym 1,2-butanediol, 1,2-butylene glycol, 1,2-dihydroxybutane, alpha-butyleneglycol, alpha-butylene glycol, 1,2-butandiol, 1,2-dihydroxy butane, butanediol, 1,2, .alpha.-butylene glycol, 1,2-butylene-glycol
PubChem CID 11429
ChEBI CHEBI:52682
IUPAC Name butane-1,2-diol
SMILES CCC(O)CO

Specifications

Color Yellow
Boiling Point 194°C
Quantity 25 g
Formula Weight 90.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Butanediol
EINECSNumber (2)-0235
RTECSNumber EK0380000
TSCA Yes