CAS RN 584-03-2
CAS RN 584-03-2
IUPAC Name:
butane-1,2-diol
Synonyms:
1,2-butanediol α-butylene glycol α-butyleneglycol 1,2-butylene glycol ethylethylene glycol 1,2-(dihydroxy)butane
Molecular Weight (g/mol):
90.12
Molecular Formula:
C4H10O2
InChi Key:
BMRWNKZVCUKKSR-UHFFFAOYNA-N
SMILES:
CCC(O)CO
1
–
1
of
1
results
1
1,2-Butanediol 98.0+%, TCI America™
CAS: 584-03-2 Molecular Formula: C4H10O2 Molecular Weight (g/mol): 90.12 MDL Number: MFCD00004570 InChI Key: BMRWNKZVCUKKSR-UHFFFAOYNA-N Synonym: 1,2-butanediol,1,2-butylene glycol,1,2-dihydroxybutane,alpha-butyleneglycol,alpha-butylene glycol,1,2-butandiol,1,2-dihydroxy butane,butanediol, 1,2,.alpha.-butylene glycol,1,2-butylene-glycol PubChem CID: 11429 ChEBI: CHEBI:52682 IUPAC Name: butane-1,2-diol SMILES: CCC(O)CO