missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethyl)naphthalene, 97%

Catalog No. p-7058908
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4349103 1 g
AA4349106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4349103 Supplier Thermo Scientific Chemicals Supplier No. 04349103
Only null left

CAS: 13686-49-2 | C12H11Br | 235.124 g/mol

1-(2-Bromoethyl)naphthalene has applications in research field such as Western blotting, Immunoprecipitation, immunogen, and in purification, process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13686-49-2
Molecular Formula C12H11Br
Molecular Weight (g/mol) 235.124
MDL Number MFCD00037737
InChI Key GPHCPUFIWQJZOI-UHFFFAOYSA-N
PubChem CID 139541
IUPAC Name 1-(2-bromoethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCBr

Specifications

Boiling Point 237°C to 238°C (1.65 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Synonym 1-2-bromoethyl naphthalene, 2-1-naphthyl ethyl bromide, naphthalene, 1-2-bromoethyl, 2-bromoethylnaphthalene, 1-2-bromoethyl naphtalene, 1-2-bromo-ethyl-naphthalene, naphthalene,1-2-bromoethyl, 2-alpha-naphthyl ethyl bromide, naphthylathylbromid, 2-bromoethyl napthalene
Solubility Information It is slightly soluble in water.
Formula Weight 235.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1- (2-Bromoethyl)naphthalene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only