missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Bromoethoxy)-4-fluorobenzene, 97+%

Catalog No. p-7034707
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5240303 1 g
AAH5240306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5240303 Supplier Thermo Scientific Chemicals Supplier No. H5240303
Only null left

CAS: 332-48-9 | C8H8BrFO | 219.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 332-48-9
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD00044739
InChI Key JXSPKRUNMHMICQ-UHFFFAOYSA-N
Synonym 1-2-bromoethoxy-4-fluorobenzene, 4-fluorophenoxy-ethylbromide, 2-bromoethyl 4-fluorophenyl ether, 4-fluorophenoxyethylbromide, 4-2-bromoethoxy fluorobenzene, 4-fluorophenoxyethyl bromide, beta-bromo-4-fluorophenetole, 2-4-fluorophenoxy ethyl bromide, benzene, 1-2-bromoethoxy-4-fluoro, p-fluorophenoxy ethylbromide
PubChem CID 2064171
IUPAC Name 1-(2-bromoethoxy)-4-fluorobenzene
SMILES FC1=CC=C(OCCBr)C=C1

Specifications

Quantity 1 g
Formula Weight 219.05
Percent Purity ≥97%
Chemical Name or Material 1-(2-Bromoethoxy)-4-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only