missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethylsilyloxy)cyclobutene 94.0+%, TCI America™

Catalog No. p-7119252
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17681G 1 g
B17685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17681G Supplier TCI America Supplier No. B17681G
Only null left

Chemical Identifiers

CAS 17082-61-0
Molecular Formula C10H22O2Si2
Molecular Weight (g/mol) 230.454
MDL Number MFCD00010798
InChI Key WOBRFSDEZREQAB-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsilyloxy cyclobutene, bis trimethylsiloxy cyclobutene, 1,2-bis trimethylsilyl oxy cyclobut-1-ene, 1,2-bis trimethylsiloxy cyclobutene, silane, 1-cyclobutene-1,2-diylbis oxy bis trimethyl, acmc-1c5q5, 1,2-bistrimethylsilyloxycyclobutene, 1,2-bis trimethylsilyloxycyclobutene, cyclobut-1-ene-1,2-diylbis oxy bis trimethylsilane
PubChem CID 2734034
IUPAC Name trimethyl-(2-trimethylsilyloxycyclobuten-1-yl)oxysilane
SMILES C[Si](C)(C)OC1=C(CC1)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 59°C
Quantity 1 g
Formula Weight 230.45
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethylsilyloxy)cyclobutene