accessibility menu, dialog, popup

UserName

CAS RN 17082-61-0

IUPAC Name: trimethyl({2-[(trimethylsilyl)oxy]cyclobut-1-en-1-yl}oxy)silane
Synonyms: trimethyl({2-[(trimethylsilyl)oxy]cyclobut-1-en-1-yl}oxy)silane
Molecular Weight (g/mol): 230.45
Molecular Formula: C10H22O2Si2
InChi Key: WOBRFSDEZREQAB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=C(CC1)O[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results