missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trimethylsilyl)benzene 95.0+%, TCI America™

Catalog No. p-7119606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22991G 1 g
B22995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22991G Supplier TCI America Supplier No. B22991G
Only null left

Chemical Identifiers

CAS 17151-09-6
Molecular Formula C12H22Si2
Molecular Weight (g/mol) 222.478
MDL Number MFCD00015590
InChI Key YHMJZIJXVNRXIN-UHFFFAOYSA-N
Synonym 1,2-bis trimethylsilyl benzene, trimethyl 2-trimethylsilyl phenyl silane, trimethyl-2-trimethylsilylphenyl silane, acmc-1c8d9, benzene,1,2-bis trimethylsilyl, 1,2-phenylenebis trimethylsilane, trimethyl 2-trimethylsilyl phenyl silane #
PubChem CID 519794
IUPAC Name trimethyl-(2-trimethylsilylphenyl)silane
SMILES C[Si](C)(C)C1=CC=CC=C1[Si](C)(C)C

Specifications

Color Colorless
Quantity 1 g
Formula Weight 222.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trimethylsilyl)benzene
TSCA No
Recommended Storage Refrigerator