missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trifluoromethyl)benzene 97.0+%, TCI America™

Catalog No. B31105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31105G 5 g
B311025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31105G Supplier TCI America Supplier No. B31105G

Chemical Identifiers

CAS 433-95-4
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD03094218
InChI Key XXZOEDQFGXTEAD-UHFFFAOYSA-N
PubChem CID 9818
IUPAC Name 1,2-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC=C1C(F)(F)F

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 1993
Synonym alpha,alpha,alpha,alpha′C,alpha′C,alpha′C-Hexafluoro-o-xylene
Formula Weight 214.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(trifluoromethyl)benzene