missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(trichlorosilyl)ethane 97.0+%, TCI America™

Catalog No. B12925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12925G 5 g
B129225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12925G Supplier TCI America Supplier No. B12925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2504-64-5
Molecular Formula C2H4Cl6Si2
Molecular Weight (g/mol) 296.924
MDL Number MFCD00013601
InChI Key WDVUXWDZTPZIIE-UHFFFAOYSA-N
PubChem CID 75631
IUPAC Name trichloro(2-trichlorosilylethyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)[Si](Cl)(Cl)Cl

Specifications

Color White
Quantity 5 g
UN Number 2987
Formula Weight 296.92
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Bis(trichlorosilyl)ethane