accessibility menu, dialog, popup

UserName

ClSiClClSiClClCl

CAS RN 2504-64-5

IUPAC Name: trichloro[2-(trichlorosilyl)ethyl]silane
Synonyms: trichloro[2-(trichlorosilyl)ethyl]silane
Molecular Weight (g/mol): 296.92
Molecular Formula: C2H4Cl6Si2
InChi Key: WDVUXWDZTPZIIE-UHFFFAOYSA-N
SMILES: Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)