missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(phenylthio)ethylene (cis- and trans- mixture) 98.0+%, TCI America™
$143.54 - $438.05
Chemical Identifiers
| CAS | 23528-44-1 |
|---|---|
| Molecular Formula | C14H12S2 |
| Molecular Weight (g/mol) | 244.37 |
| InChI Key | UHGSEDVQATYWGI-VAWYXSNFSA-N |
| Synonym | 1,2-Bis(phenylmercapto)ethylene |
| PubChem CID | 5372209 |
| IUPAC Name | [(E)-2-phenylsulfanylethenyl]sulfanylbenzene |
| SMILES | C1=CC=C(C=C1)SC=CSC2=CC=CC=C2 |
Chemical Identifiers
| 23528-44-1 | |
| 244.37 | |
| 1,2-Bis(phenylmercapto)ethylene | |
| [(E)-2-phenylsulfanylethenyl]sulfanylbenzene |
| C14H12S2 | |
| UHGSEDVQATYWGI-VAWYXSNFSA-N | |
| 5372209 | |
| C1=CC=C(C=C1)SC=CSC2=CC=CC=C2 |
Specifications
| 23528-44-1 | |
| 166°C | |
| 5 g | |
| UHGSEDVQATYWGI-VAWYXSNFSA-N | |
| [(E)-2-phenylsulfanylethenyl]sulfanylbenzene | |
| 5372209 | |
| ≥98.0% (GC) | |
| 1,2-Bis(phenylthio)ethylene (cis- and trans- mixture) |
| 48°C | |
| C14H12S2 | |
| 1,2-Bis(phenylmercapto)ethylene | |
| C1=CC=C(C=C1)SC=CSC2=CC=CC=C2 | |
| 244.37 | |
| 244.37 | |
| Crystals |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator