missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylthio)ethane 98.0+%, TCI America™

Catalog No. B33535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33535G Supplier TCI America Supplier No. B33535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 622-20-8
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.39
MDL Number MFCD00014085
InChI Key MHCVYAFXPIMYRD-UHFFFAOYSA-N
Synonym 1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
PubChem CID 69317
IUPAC Name {[2-(phenylsulfanyl)ethyl]sulfanyl}benzene
SMILES C(CSC1=CC=CC=C1)SC1=CC=CC=C1

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 65°C
Quantity 5 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(phenylthio)ethane