missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylthio)ethane, 98+%

Catalog No. AAL0201909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0201909 10 g
AAL0201918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0201909 Supplier Thermo Scientific Chemicals Supplier No. L0201909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 622-20-8 | C14H14S2 | 246.39 g/mol

1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 622-20-8
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.39
MDL Number MFCD00014085
InChI Key MHCVYAFXPIMYRD-UHFFFAOYSA-N
Synonym 1,2-bis phenylthio ethane, ethane, 1,2-bis phenylthio, bis phenylthio ethane, 1,1'-ethane-1,2-diylbis thio dibenzene, 2-phenylsulfanyl ethyl sulfanyl benzene, benzene, 1,1'-1,2-ethanediylbis thio bis, 1,1'-1,2-ethanediylbis thio bis-benzene, 1,1'-ethane-1,2-diylbis thio bisbenzene, 2-phenylthioethylthio benzene
PubChem CID 69317
IUPAC Name 2-phenylsulfanylethylsulfanylbenzene
SMILES C(CSC1=CC=CC=C1)SC1=CC=CC=C1

Specifications

Melting Point 68°C to 70°C
Linear Formula C6H5SCH2CH2SC6H5
Quantity 10 g
Beilstein 1877719
Solubility Information Insoluble in water.
Formula Weight 246.39
Percent Purity ≥98%
Chemical Name or Material 1,2-Bis(phenylthio)ethane
EINECSNumber 210-723-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only