missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate 96.0+%, TCI America™

Catalog No. B3292200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3292200MG 200 mg
B32921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3292200MG Supplier TCI America Supplier No. B3292200MG

Chemical Identifiers

CAS 858971-43-4
Molecular Formula C18H20O6PdS2
Molecular Weight (g/mol) 502.892
MDL Number MFCD09842752
InChI Key SNNYSJNYZJXIFE-UHFFFAOYSA-L
Synonym White Catalyst
PubChem CID 11562441
IUPAC Name 2-(benzenesulfinyl)ethylsulfinylbenzene;palladium(2+);diacetate
SMILES CC(=O)[O-].CC(=O)[O-].C1=CC=C(C=C1)S(=O)CCS(=O)C2=CC=CC=C2.[Pd+2]

Specifications

Quantity 200 mg
Formula Weight 502.89
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(phenylsulfinyl)ethane Palladium(II) Diacetate
TSCA No
Recommended Storage Freezer