missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(methanesulfonamido)benzene, 97%
CAS: 7596-80-7 | C8H12N2O4S2 | 264.31 g/mol
Supplier: Thermo Scientific Chemicals A1983009
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,2-Bis(methanesulfonamido)benzene | |
| 211°C to 214°C | |
| C8H12N2O4S2 | |
| 10 g | |
| BYZJRLRCNOECEV-UHFFFAOYSA-N | |
| N-[2-(methanesulfonamido)phenyl]methanesulfonamide | |
| 227307 | |
| 97% |
| 7596-80-7 | |
| Odorless | |
| MFCD00276605 | |
| 1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide | |
| CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O | |
| 264.31 | |
| 264.32 |
Chemical Identifiers
| 7596-80-7 | |
| 264.31 | |
| BYZJRLRCNOECEV-UHFFFAOYSA-N | |
| 227307 | |
| CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O |
| C8H12N2O4S2 | |
| MFCD00276605 | |
| 1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide | |
| N-[2-(methanesulfonamido)phenyl]methanesulfonamide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only