missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(methanesulfonamido)benzene, 97%

Catalog No. AAA1983088
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1983088 2.5 g
AAA1983009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1983088 Supplier Thermo Scientific Chemicals Supplier No. A1983088
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7596-80-7 | C8H12N2O4S2 | 264.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7596-80-7
Molecular Formula C8H12N2O4S2
Molecular Weight (g/mol) 264.31
MDL Number MFCD00276605
InChI Key BYZJRLRCNOECEV-UHFFFAOYSA-N
Synonym 1,2-bis methanesulfonamido benzene, n,n'-1,2-phenylene dimethanesulfonamide, n-2-methanesulfonamidophenyl methanesulfonamide, n-2-methylsulfonyl amino phenyl methanesulfonamide, methylsulfonyl 2-methylsulfonyl amino phenyl amine, maybridge1_000065, n,n'-o-phenylenebis methanesulfonamide, o-phenylenediamine n,n'-bis methanesulfonamide, n-2-methanesulfonamido phenyl methanesulfonamide
PubChem CID 227307
IUPAC Name N-[2-(methanesulfonamido)phenyl]methanesulfonamide
SMILES CS(=O)(=O)NC1=CC=CC=C1NS(C)(=O)=O

Specifications

Melting Point 211°C to 214°C
Odor Odorless
Quantity 2.5 g
Formula Weight 264.32
Percent Purity 97%
Chemical Name or Material 1,2-Bis(methanesulfonamido)benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only