missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(diphenylphosphino)benzene 98.0+%, TCI America™
$169.41 - $508.25
Chemical Identifiers
| CAS | 13991-08-7 |
|---|---|
| Molecular Formula | C30H24P2 |
| Molecular Weight (g/mol) | 446.47 |
| MDL Number | MFCD00014081 |
| InChI Key | NFRYVRNCDXULEX-UHFFFAOYSA-N |
| Synonym | 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene |
| PubChem CID | 498379 |
| IUPAC Name | [2-(diphenylphosphanyl)phenyl]diphenylphosphane |
| SMILES | C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 13991-08-7 | |
| 446.47 | |
| NFRYVRNCDXULEX-UHFFFAOYSA-N | |
| 498379 | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C30H24P2 | |
| MFCD00014081 | |
| 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene | |
| [2-(diphenylphosphanyl)phenyl]diphenylphosphane |
Specifications
| 13991-08-7 | |
| White-Yellow | |
| MFCD00014081 | |
| 1,2-bis diphenylphosphino benzene, dppbz, dppbe, dppben, dppbenz, o-bis diphenylphosphino benzene, 1,2-bis dimethylphosphino benzene, o-phenylenebis diphenylphosphine, phosphine, phenylenebis diphenyl, 1,2-bis diphenylphosphanyl benzene | |
| C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 446.47 | |
| 446.47 | |
| Crystalline Powder |
| 188°C | |
| C30H24P2 | |
| 1 g | |
| NFRYVRNCDXULEX-UHFFFAOYSA-N | |
| [2-(diphenylphosphanyl)phenyl]diphenylphosphane | |
| 498379 | |
| ≥98.0% (GC) | |
| 1,2-Bis(diphenylphosphino)benzene |
Safety and Handling
TSCA : No