missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylsilyl)benzene 94.0+%, TCI America™

Catalog No. B16995ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16995ML 5 mL
B16991ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16995ML Supplier TCI America Supplier No. B16995ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17985-72-7
Molecular Formula C10H16Si2
Molecular Weight (g/mol) 192.408
MDL Number MFCD00142462
InChI Key MUUXBTFQEXVEEI-UHFFFAOYSA-N
Synonym 1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
PubChem CID 6364986
IUPAC Name [2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC=CC=C1[Si](C)C

Specifications

Color Yellow
Boiling Point 128°C
Quantity 5 mL
Formula Weight 194.42
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(dimethylsilyl)benzene