accessibility menu, dialog, popup

UserName

H3CSiCH3SiCH3H3C

CAS RN 17985-72-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 192.41
Molecular Formula: C10H16Si2
InChi Key: MUUXBTFQEXVEEI-UHFFFAOYSA-N
SMILES: C[Si](C)C1=CC=CC=C1[Si](C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)