missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethylsilyl)benzene, 98%

Catalog No. AA4375504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4375504 2 g
AA4375509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4375504 Supplier Thermo Scientific Chemicals Supplier No. 04375504

CAS: 17985-72-7 | C10H16Si2 | 192.408 g/mol

1,2-Bis(dimethylsilyl)benzene is a protecting group-reagent for amines and amino acids forming benzostabase derivatives. It is an important raw material and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17985-72-7
Molecular Formula C10H16Si2
Molecular Weight (g/mol) 192.408
MDL Number MFCD00142462
InChI Key MUUXBTFQEXVEEI-UHFFFAOYSA-N
Synonym 1,2-bis dimethylsilyl benzene, 1,2-phenylenebis dimethylsilane, 2-dimethyl, acmc-209efy, bis dimethylsilyl benzene, amtsi084, o-phenylenebis dimethylsilane, 2-dimethylsilylphenyl-dimethylsilane, 5,6-bis dimethylsilanediyl-1,3-cyclohexadiene
PubChem CID 6364986
IUPAC Name [2-(dimethyl-$l^{3}-silanyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC=CC=C1[Si](C)C

Specifications

Density 0.898 g/mL
Boiling Point 101°C (13 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.514
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 194.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis (dimethylsilyl)benzene
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only