missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(dimethoxyphosphoryl)benzene, 99%

Catalog No. AA3927304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3927304 2 g
AA3927309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3927304 Supplier Thermo Scientific Chemicals Supplier No. 03927304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 15104-46-8 | C10H16O6P2 | 294.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15104-46-8
Molecular Formula C10H16O6P2
Molecular Weight (g/mol) 294.18
MDL Number MFCD00142461
InChI Key TUKTVDDATWNXSN-UHFFFAOYSA-N
Synonym 1,2-bis dimethoxyphosphoryl benzene, tetramethyl 1,2-phenylenebis phosphonate, tetramethyl-1,2-phenylenediphosphonate, dimethyl 2-dimethoxyphosphoryl phenylphosphonate, acmc-20ap6x, bis dimethoxyphosphoryl benzene, 1,2-bis dimethoxyphosphinyl benzene, 1,2-bis dimethoxyphosphoryl benzene 500mg, 2-dimethoxycarbonyl phenyl dimethoxyphosphino-1-one
PubChem CID 2733153
IUPAC Name 1,2-bis(dimethoxyphosphoryl)benzene
SMILES COP(=O)(C1=CC=CC=C1P(=O)(OC)OC)OC

Specifications

Melting Point 80°C to 82°C
Quantity 2 g
UN Number UN3464
Formula Weight 294.18
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material 1,2-Bis(dimethoxyphosphoryl)benzene
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only