missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(5-methyl-2-benzoxazolyl)ethylene 98.0+%, TCI America™

Catalog No. b425025g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B425025G 25 g
B42505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B425025G Supplier TCI America Supplier No. B425025G
Only null left

Chemical Identifiers

CAS 1041-00-5
Molecular Formula C18H14N2O2
Molecular Weight (g/mol) 290.322
MDL Number MFCD00408425
InChI Key VKRZNAWSCAUDRQ-BQYQJAHWSA-N
PubChem CID 5332173
IUPAC Name 5-methyl-2-[(E)-2-(5-methyl-1,3-benzoxazol-2-yl)ethenyl]-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C=CC3=NC4=C(O3)C=CC(=C4)C

Specifications

Melting Point 189°C
Color Yellow
Quantity 25 g
Formula Weight 290.32
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(5-methyl-2-benzoxazolyl)ethylene
EINECSNumber (5)-0165&(5)-2710
TSCA Yes