missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate 98.0+%, TCI America™

$182.68 - $631.92

Chemical Identifiers

CAS 196599-80-1
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
InChI Key MTNUOOJIYAKURM-UHFFFAOYSA-N
Synonym Cyclohexylcarbamic Acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl Ester
PubChem CID 10596958
IUPAC Name [1,2-bis(4-methoxyphenyl)-2-oxoethyl] N-cyclohexylcarbamate
SMILES COC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)OC)OC(=O)NC3CCCCC3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5085200MG
View Documents
TCI America
B5085200MG
200 mg
Each for $182.68
Only null left
 
B50851G
View Documents
TCI America
B50851G
1 g
Each for $631.92
Only null left
 
Description

Description

Specifications

Chemical Identifiers

196599-80-1
397.471
Cyclohexylcarbamic Acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl Ester
[1,2-bis(4-methoxyphenyl)-2-oxoethyl] N-cyclohexylcarbamate
C23H27NO5
MTNUOOJIYAKURM-UHFFFAOYSA-N
10596958
COC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)OC)OC(=O)NC3CCCCC3

Specifications

196599-80-1
White
200 mg
MTNUOOJIYAKURM-UHFFFAOYSA-N
[1,2-bis(4-methoxyphenyl)-2-oxoethyl] N-cyclohexylcarbamate
10596958
≥98.0% (HPLC,N)
1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate
118°C
C23H27NO5
Cyclohexylcarbamic Acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl Ester
COC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)OC)OC(=O)NC3CCCCC3
397.471
397.47
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'