missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate 98.0+%, TCI America™

Catalog No. B50851G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50851G 1 g
B5085200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50851G Supplier TCI America Supplier No. B50851G
Only null left

Chemical Identifiers

CAS 196599-80-1
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
InChI Key MTNUOOJIYAKURM-UHFFFAOYSA-N
Synonym Cyclohexylcarbamic Acid 1,2-Bis(4-methoxyphenyl)-2-oxoethyl Ester
PubChem CID 10596958
IUPAC Name [1,2-bis(4-methoxyphenyl)-2-oxoethyl] N-cyclohexylcarbamate
SMILES COC1=CC=C(C=C1)C(C(=O)C2=CC=C(C=C2)OC)OC(=O)NC3CCCCC3

Specifications

Melting Point 118°C
Color White
Quantity 1 g
Formula Weight 397.47
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(4-methoxyphenyl)-2-oxoethyl Cyclohexylcarbamate