Learn More
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%
CAS: 136735-95-0 | C18H28P2 | 306.37 g/mol
Supplier: Thermo Scientific Chemicals H6064703
Description
It is a DuPhos and BPE ligands which are highly efficient privileged ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a DuPhos and BPE ligands which are highly efficient privileged ligands.
Solubility
Insoluble in water.
Notes
Air sensitive. Store at room temperature.
Specifications
| 1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene | |
| 66°C to 76°C | |
| MFCD00142322 | |
| 4810602 | |
| s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane | |
| AJNZWRKTWQLAJK-VGWMRTNUSA-N | |
| (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane | |
| 2734553 | |
| ≥97% |
| 136735-95-0 | |
| C18H28P2 | |
| 1 g | |
| Air Sensitive | |
| Insoluble in water. | |
| CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C | |
| 306.37 | |
| 306.36 |
Chemical Identifiers
| 136735-95-0 | |
| 306.37 | |
| AJNZWRKTWQLAJK-VGWMRTNUSA-N | |
| 2734553 | |
| CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C |
| C18H28P2 | |
| MFCD00142322 | |
| s,s-me-duphos, methyl-duphos, s,s, s,s-methyl-duphos, unii-hl745j99sb, +-1,2-bis 2s,5s-2,5-dimethylphospholano benzene, phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's, s,s-me-duphos mi, s,s-methyl-duphos, +, 2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane | |
| (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only