CAS RN 136735-95-0
IUPAC Name:
1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Synonyms:
1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol):
306.37
Molecular Formula:
C18H28P2
InChi Key:
AJNZWRKTWQLAJK-UHFFFAOYNA-N
SMILES:
CC1CCC(C)P1C1=CC=CC=C1P1C(C)CCC1C
1
–
1
of
1
results
1
Promotions Available
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%
1,2-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]benzene, 97+%
CAS: 136735-95-0 Molecular Formula: C18H28P2 Molecular Weight (g/mol): 306.37 MDL Number: MFCD00142322 InChI Key: AJNZWRKTWQLAJK-VGWMRTNUSA-N Synonym: s,s-me-duphos,methyl-duphos, s,s,s,s-methyl-duphos,unii-hl745j99sb,+-1,2-bis 2s,5s-2,5-dimethylphospholano benzene,phospholane, 1,1'-1,2-phenylene bis 2,5-dimethyl-, 2s,2's,5s,5's,s,s-me-duphos mi,s,s-methyl-duphos, +,2s,5s-1-2-2s,5s-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane PubChem CID: 2734553 IUPAC Name: (2S,5S)-1-[2-[(2S,5S)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethylphospholane SMILES: CC1CCC(P1C2=CC=CC=C2P3C(CCC3C)C)C