missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-iodoethoxy)ethane (stabilized with Copper chip) 96.0+%, TCI America™

Catalog No. B168710G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B168710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B168710G Supplier TCI America Supplier No. B168710G

Chemical Identifiers

CAS 36839-55-1
Molecular Formula C6H12I2O2
Molecular Weight (g/mol) 369.969
MDL Number MFCD00075008
InChI Key BCAGFJXMCZSAHD-UHFFFAOYSA-N
Synonym 1,2-bis 2-iodoethoxy ethane, ethane,1,2-bis 2-iodoethoxy, 1-iodo-2-2-2-iodoethoxy ethoxy ethane, diiodine-peg3, acmc-209tta, 1,2-bis 2-iodoethoxy-ethane, 1,2-bis-2-iodoethoxy ethane, 1,2-bis 2-iodanylethoxy ethane, 1,2-bis-2-iodoethyloxy ethane, ethane, 1,2-bis 2-iodoethoxy
PubChem CID 593436
IUPAC Name 1,2-bis(2-iodoethoxy)ethane
SMILES C(COCCI)OCCI

Specifications

Quantity 10 g
Formula Weight 369.97
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Bis(2-iodoethoxy)ethane (stabilized with Copper chip)