accessibility menu, dialog, popup

UserName

OIOI

CAS RN 36839-55-1

IUPAC Name: 1-iodo-2-[2-(2-iodoethoxy)ethoxy]ethane
Synonyms: 1-iodo-2-[2-(2-iodoethoxy)ethoxy]ethane
Molecular Weight (g/mol): 369.97
Molecular Formula: C6H12I2O2
InChi Key: BCAGFJXMCZSAHD-UHFFFAOYSA-N
SMILES: ICCOCCOCCI
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)