missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid 90.0+%, TCI America™

Catalog No. B3895100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3895100MG 100 mg
B38951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3895100MG Supplier TCI America Supplier No. B3895100MG

Chemical Identifiers

CAS 85233-19-8
Molecular Formula C22H24N2O10
Molecular Weight (g/mol) 476.44
MDL Number MFCD00036255
InChI Key FTEDXVNDVHYDQW-UHFFFAOYSA-N
Synonym bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid
PubChem CID 104751
ChEBI CHEBI:60888
IUPAC Name 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
SMILES OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O

Specifications

Melting Point 159°C
Color White
Quantity 100 mg
Formula Weight 476.44
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic Acid
RTECSNumber MC0423000
TSCA No
Recommended Storage Refrigerator