missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 352°C |
| Quantity | 500 g |
| Synonym | 2,2′,3,3′,4,4′,5,5′,6,6′-Decabromobibenzyl |
| Formula Weight | 971.23 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,2-Bis(2,3,4,5,6-pentabromophenyl)ethane |
Safety and Handling
| RTECSNumber | DA0358200 |
| TSCA | Yes |