missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzisoxazole 96.0+%, TCI America™
Supplier: TCI America B096925G
Specifications
| 1,2-Benzisoxazole | |
| Yellow | |
| C7H5NO | |
| 25 g | |
| KTZQTRPPVKQPFO-UHFFFAOYSA-N | |
| 1,2-benzoxazole | |
| 71073 | |
| 119.12 | |
| Liquid |
| 271-95-4 | |
| 100°C | |
| MFCD00005852 | |
| 1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine | |
| C1=CC=C2C(=C1)C=NO2 | |
| 119.123 | |
| CHEBI:51554 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 271-95-4 | |
| 119.123 | |
| KTZQTRPPVKQPFO-UHFFFAOYSA-N | |
| 71073 | |
| 1,2-benzoxazole |
| C7H5NO | |
| MFCD00005852 | |
| 1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine | |
| CHEBI:51554 | |
| C1=CC=C2C(=C1)C=NO2 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator