missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisoxazole 96.0+%, TCI America™

Catalog No. p-7118772
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09695G 5 g
B096925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09695G Supplier TCI America Supplier No. B09695G
Only null left

Chemical Identifiers

CAS 271-95-4
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.123
MDL Number MFCD00005852
InChI Key KTZQTRPPVKQPFO-UHFFFAOYSA-N
Synonym 1,2-benzisoxazole, benzo d isoxazole, indoxazene, benzisoxazole, benz d isoxazole, 4,5-benzisoxazole, 1-oxa-2-aza-1h-indene, benzoisoxazole, indoxazine, isoindoxazine
PubChem CID 71073
ChEBI CHEBI:51554
IUPAC Name 1,2-benzoxazole
SMILES C1=CC=C2C(=C1)C=NO2

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
Formula Weight 119.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Benzisoxazole
TSCA No
Recommended Storage Refrigerator