missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzenedimethanol, 97%

Catalog No. p-3777384
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180410025 2.5 g Glass bottle
AC180410100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC180410025 Supplier Thermo Scientific Chemicals Supplier No. 180410025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-14-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-14-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004626
InChI Key XMUZQOKACOLCSS-UHFFFAOYSA-N
Synonym 1,2-benzenedimethanol, 1,2-phenylenedimethanol, phthalyl alcohol, 1,2-dihydroxymethylbenzene, o-xylene-alpha,alpha'-diol, 2-hydroxymethyl phenyl methanol, benzenedimethanol, o-xylylene glycol, 1,2-di hydroxymethyl benzene, 2-hydroxymethylbenzenemethanol
PubChem CID 69153
IUPAC Name [2-(hydroxymethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White to Yellow
Boiling Point 291.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CH2OH)2
Quantity 2.5 g
Beilstein 06, 910
Fieser 16,18; 12,563; 13,341
Solubility Information Solubility in water: soluble
Formula Weight 138.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 2-Benzenedimethanol
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-293-9

RUO – Research Use Only