missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193433
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019942 5 mL
111018188 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019942 Supplier Merck Emd Millipore Supplier No. 956805ML

p-Xylene is an aromatic liquid, which is an isomeric form of xylene. It can be produced via Diels-Alder reaction between 2,5-dimethylfuran and ethylene catalyzed by zeolite.

Specifications

Packaging Glass Bottle
CAS 106-42-3
Vapor Pressure 9 mmHg (20°C)
Vapor Density 3.7 (vs. air)
Density 0.861 g/mL (at 20°C)
Physical Form Neat
Linear Formula C6H4(CH3)2
MDL Number MFCD00008556
Melting Point 12°C to 13°C
Boiling Point 138°C
Synonym 1,4-Dimethylbenzene
Quantity 5 mL
Formula Weight 106.17
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less