missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol 99.0+%, TCI America™

Supplier:  TCI America H106925G

Quantity 25 g

 View more versions of this product

Catalog No. H106925G


Only null left

Description

Description

Specifications

Chemical Identifiers

106-69-4
134.18
ZWVMLYRJXORSEP-UHFFFAOYNA-N
7823
OCCCCC(O)CO
C6H14O3
MFCD00002976
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
hexane-1,2,6-triol

Specifications

1,2,6-Hexanetriol
White
178°C
25 g
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
OCCCCC(O)CO
134.18
134.18
Crystalline Powder
106-69-4
4.5 to 7.5 (300g/L H2O 20°C)
C6H14O3
MFCD00002976
ZWVMLYRJXORSEP-UHFFFAOYNA-N
hexane-1,2,6-triol
7823
≥99.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0244

RTECSNumber : MO4650000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions