missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol 99.0+%, TCI America™

Catalog No. p-7130889
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10695G 5 g
H106925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10695G Supplier TCI America Supplier No. H10695G
Only null left

Chemical Identifiers

CAS 106-69-4
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002976
InChI Key ZWVMLYRJXORSEP-UHFFFAOYNA-N
Synonym 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
PubChem CID 7823
IUPAC Name hexane-1,2,6-triol
SMILES OCCCCC(O)CO

Specifications

Color White
pH 4.5 to 7.5 (300g/L H2O 20°C)
Boiling Point 178°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,6-Hexanetriol
EINECSNumber (2)-0244
RTECSNumber MO4650000
TSCA Yes
Recommended Storage Refrigerator