missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Dimethylphenoxy)-2-propanone 95.0+%, TCI America™

$87.60 - $262.83

Chemical Identifiers

CAS 53012-41-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00130266
InChI Key XDJULAUHYAJQBU-UHFFFAOYSA-N
Synonym (2,6-Dimethylphenoxy)acetone
PubChem CID 104378
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-one
SMILES CC1=C(C(=CC=C1)C)OCC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D51211G
View Documents
TCI America
D51211G
1 g
Each for $87.60
Only null left
 
D51215G
View Documents
TCI America
D51215G
5 g
Each for $262.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

53012-41-2
178.231
XDJULAUHYAJQBU-UHFFFAOYSA-N
104378
CC1=C(C(=CC=C1)C)OCC(=O)C
C11H14O2
MFCD00130266
(2,6-Dimethylphenoxy)acetone
1-(2,6-dimethylphenoxy)propan-2-one

Specifications

53012-41-2
113°C
Clear Liquid at 20°C
MFCD00130266
XDJULAUHYAJQBU-UHFFFAOYSA-N
1-(2,6-dimethylphenoxy)propan-2-one
104378
≥95.0% (GC)
Yellow
1 g
C11H14O2
(2,6-Dimethylphenoxy)acetone
CC1=C(C(=CC=C1)C)OCC(=O)C
178.231
178.23
1-(2,6-Dimethylphenoxy)-2-propanone
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'