missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2,6-Dimethylphenoxy)-2-propanone 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Physical Form | Clear Liquid at 20°C |
| Boiling Point | 113°C |
| Formula Weight | 178.23 |
| Percent Purity | ≥95.0% (GC) |
| Chemical Name or Material | 1-(2,6-Dimethylphenoxy)-2-propanone |