missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Diisopropylphenyl)imidazole 98.0+%, TCI America™

Catalog No. D49481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49481G 1 g
D49485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49481G Supplier TCI America Supplier No. D49481G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25364-47-0
Molecular Formula C15H20N2
Molecular Weight (g/mol) 228.339
MDL Number MFCD13299721
InChI Key OWSXDWUAPKBLOI-UHFFFAOYSA-N
PubChem CID 15879265
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]imidazole
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=CN=C2

Specifications

Melting Point 130°C
Color White
Boiling Point 105°C
Quantity 1 g
Formula Weight 228.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,6-Diisopropylphenyl)imidazole